2-(4-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
Supplier: Matrix Scientific
049122-2.5G
049122-500MG
101886-394EA
838.46
USD
101886-394
101886-392
2-(4-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
2-(4-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClNO₃ |
MDL Number:
MFCD13559685 CAS Number: 1219949-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...