N-(2-Furoyl)-β-alanine
Supplier: Matrix Scientific
049116-2.5G
049116-500MG
101886-370EA
838.46
USD
101886-370
101886-368
N-(2-Furoyl)-β-alanine
N-(2-Furoyl)-beta-alanine
Hazard: Irritant
Formula:
C₈H₉NO₄ MW: 183.17 g/mol |
MDL Number:
MFCD00466494 CAS Number: 5652-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...