1-(2-Furoyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049112-500MG 049112-2.5G
101886-352EA 364.62 USD
101886-352 101886-354
1-(2-Furoyl)-2-piperidinecarboxylic acid
1-(2-Furoyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD08444976
CAS Number: 1025450-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR