N-[(4-Chloro-2-pyridinyl)carbonyl]-β-alanine

Supplier: Matrix Scientific

049098-500MG 049098-2.5G
101886-296EA 364.62 USD
101886-296 101886-298
N-[(4-Chloro-2-pyridinyl)carbonyl]-β-alanine
N-[(4-Chloro-2-pyridinyl)carbonyl]-beta-alanine

Hazard: Irritant

Formula: C₉H₉ClN₂O₃
MW: 228.64 g/mol
MDL Number: MFCD12094725
CAS Number: 1154331-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR