2-(3-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride

049083-2.5G
101886-238EA 838.46 USD
101886-238
2-(3-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride
2-(3-Piperidinyl)ethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₂O₂
MDL Number: MFCD13559659
CAS Number: 1220031-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR