4-Piperidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride

049081-2.5G
101886-230EA 838.46 USD
101886-230
4-Piperidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride
4-Piperidinylmethyl-4-chloro-2-pyridinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆Cl₂N₂O₂
MW: 291.18 g/mol
MDL Number: MFCD13559657
CAS Number: 1220020-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR