1-Isonicotinoyl-3-piperidinecarboxylic acid
Supplier: Matrix Scientific
049073-2.5G
049073-500MG
101886-198EA
838.46
USD
101886-198
101886-196
1-Isonicotinoyl-3-piperidinecarboxylic acid
1-Isonicotinoyl-3-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₃ |
MDL Number:
MFCD08444920 CAS Number: 67691-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...