1-Isonicotinoylproline

Supplier: Matrix Scientific

049071-2.5G 049071-500MG
101886-190EA 1265.91 USD
101886-190 101886-188
1-Isonicotinoylproline
1-Isonicotinoylproline

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD00448260
CAS Number: 59834-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR