4-Piperidinylmethyl isonicotinate hydrochloride

Supplier: Matrix Scientific

049061-2.5G 049061-500MG
101886-150EA 838.46 USD
101886-150 101886-148
4-Piperidinylmethyl isonicotinate hydrochloride
4-Piperidinylmethyl isonicotinate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD13559644
CAS Number: 1220031-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR