N-Acetyl-2-methylalanine

Supplier: Matrix Scientific

049054-500MG 049054-2.5G
101886-120EA 364.62 USD
101886-120 101886-122
N-Acetyl-2-methylalanine
N-Acetyl-2-methylalanine

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD13559639
CAS Number: 5362-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR