1-Acetyl-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049053-500MG
101886-116EA 320 USD
101886-116
1-Acetyl-2-piperidinecarboxylic acid
1-Acetyl-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD09044120
CAS Number: 35677-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR