2-(2-Piperidinyl)ethyl acetate hydrochloride

049046-2.5G
101886-090EA 668.58 USD
101886-090
2-(2-Piperidinyl)ethyl acetate hydrochloride
2-(2-Piperidinyl)ethyl acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD13559635
CAS Number: 1220020-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR