N-(4-Amino-2-methoxyphenyl)-2-(3-fluorophenoxy)acetamide

Supplier: Matrix Scientific

049037-500MG
101886-052EA 364.62 USD
101886-052
N-(4-Amino-2-methoxyphenyl)-2-(3-fluorophenoxy)acetamide
N-(4-Amino-2-methoxyphenyl)-2-(3-fluorophenoxy)acetamide

Hazard: Irritant

Formula: C₁₅H₁₅FN₂O₃
MW: 290.3 g/mol
MDL Number: MFCD09735855
CAS Number: 953752-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR