N-(5-Amino-2-methylphenyl)-2-(3-fluorophenoxy)acetamide

Supplier: Matrix Scientific

049028-2.5G 049028-500MG
101886-018EA 838.46 USD
101886-018 101886-016
N-(5-Amino-2-methylphenyl)-2-(3-fluorophenoxy)acetamide
N-(5-Amino-2-methylphenyl)-2-(3-fluorophenoxy)acetamide

Hazard: Irritant

Formula: C₁₅H₁₅FN₂O₂
MW: 274.3 g/mol
MDL Number: MFCD09741211
CAS Number: 954566-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR