N-(4-Amino-2-methoxyphenyl)-2-phenoxyacetamide

Supplier: Matrix Scientific

049024-500MG 049024-2.5G
101886-000EA 364.62 USD
101886-000 101886-002
N-(4-Amino-2-methoxyphenyl)-2-phenoxyacetamide
N-(4-Amino-2-methoxyphenyl)-2-phenoxyacetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MDL Number: MFCD03906889
CAS Number: 861409-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR