N-(4-Amino-2-chlorophenyl)tetrahydro-2-furancarboxamide
Supplier: Matrix Scientific
049020-2.5G
049020-500MG
101885-984EA
364.62
USD
101885-984
101885-986
N-(4-Amino-2-chlorophenyl)tetrahydro-2-furancarboxamide
N-(4-Amino-2-chlorophenyl)tetrahydro-2-furancarboxamide
Hazard: Irritant
Formula:
C₁₁H₁₃ClN₂O₂ |
MDL Number:
MFCD09044123 CAS Number: 926228-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...