N-(4-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide

Supplier: Matrix Scientific

049018-500MG 049018-2.5G
101885-976EA 364.62 USD
101885-976 101885-978
N-(4-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide
N-(4-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD09047982
CAS Number: 926272-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR