N-(5-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide
Supplier: Matrix Scientific
049014-2.5G
049014-500MG
101885-960EA
364.62
USD
101885-960
101885-962
N-(5-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide
N-(5-Amino-2-methoxyphenyl)tetrahydro-2-furancarboxamide
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ |
MDL Number:
MFCD09048726 CAS Number: 926249-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...