N-(5-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide
Supplier: Matrix Scientific
049010-2.5G
049010-500MG
101885-944EA
364.62
USD
101885-944
101885-946
N-(5-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide
N-(5-Amino-2-methylphenyl)tetrahydro-2-furancarboxamide
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD09044307 CAS Number: 926197-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...