N-(4-Amino-2-chlorophenyl)nicotinamide

Supplier: Matrix Scientific

049007-500MG 049007-2.5G
101885-932EA 364.62 USD
101885-932 101885-934
N-(4-Amino-2-chlorophenyl)nicotinamide
N-(4-Amino-2-chlorophenyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD09049740
CAS Number: 926187-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR