N-(4-Amino-2-chlorophenyl)-4-fluorobenzamide

Supplier: Matrix Scientific

048994-500MG 048994-2.5G
101885-884EA 364.62 USD
101885-884 101885-886
N-(4-Amino-2-chlorophenyl)-4-fluorobenzamide
N-(4-Amino-2-chlorophenyl)-4-fluorobenzamide

Hazard: Irritant

Formula: C₁₃H₁₀ClFN₂O
MDL Number: MFCD09044169
CAS Number: 926214-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR