N-(3-Amino-4-fluorophenyl)-4-fluorobenzamide

Supplier: Matrix Scientific

048991-500MG 048991-2.5G
101885-872EA 364.62 USD
101885-872 101885-874
N-(3-Amino-4-fluorophenyl)-4-fluorobenzamide
N-(3-Amino-4-fluorophenyl)-4-fluorobenzamide

Hazard: Irritant

Formula: C₁₃H₁₀F₂N₂O
MDL Number: MFCD09044908
CAS Number: 926219-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR