N-(3-Amino-4-methylphenyl)benzamide
Supplier: Matrix Scientific
048985-2.5G
048985-500MG
101885-850EA
838.46
USD
101885-850
101885-848
N-(3-Amino-4-methylphenyl)benzamide
N-(3-Amino-4-methylphenyl)benzamide
Hazard: Irritant
Formula:
C₁₄H₁₄N₂O |
MDL Number:
MFCD02046983 CAS Number: 221875-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...