N-(4-Amino-2-chlorophenyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

048984-2.5G 048984-500MG
101885-844EA 364.62 USD
101885-844 101885-846
N-(4-Amino-2-chlorophenyl)cyclopentanecarboxamide
N-(4-Amino-2-chlorophenyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O
MDL Number: MFCD09043305
CAS Number: 926267-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR