N-(4-Amino-2-methylphenyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

048982-2.5G 048982-500MG
101885-836EA 364.62 USD
101885-836 101885-838
N-(4-Amino-2-methylphenyl)cyclopentanecarboxamide
N-(4-Amino-2-methylphenyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD09048141
CAS Number: 926227-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR