N-(4-Aminophenyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

048981-2.5G 048981-500MG
101885-832EA 364.62 USD
101885-832 101885-834
N-(4-Aminophenyl)cyclopentanecarboxamide
N-(4-Aminophenyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD09044454
CAS Number: 926232-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR