N-(5-Amino-2-fluorophenyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

048976-2.5G 048976-500MG
101885-812EA 364.62 USD
101885-812 101885-814
N-(5-Amino-2-fluorophenyl)cyclopentanecarboxamide
N-(5-Amino-2-fluorophenyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₅FN₂O
MDL Number: MFCD09044999
CAS Number: 926250-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR