N-(4-Amino-2-chlorophenyl)cyclobutanecarboxamide
Supplier: Matrix Scientific
048971-500MG
048971-2.5G
101885-794EA
838.46
USD
101885-794
101885-792
N-(4-Amino-2-chlorophenyl)cyclobutanecarboxamide
N-(4-Amino-2-chlorophenyl)cyclobutanecarboxamide
Hazard: Irritant
Formula:
C₁₁H₁₃ClN₂O |
MDL Number:
MFCD09048718 CAS Number: 926249-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...