N-(4-Aminophenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048968-500MG 048968-2.5G
101885-780EA 364.62 USD
101885-780 101885-782
N-(4-Aminophenyl)cyclobutanecarboxamide
N-(4-Aminophenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD09043834
CAS Number: 299440-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR