N-(5-Amino-2-methoxyphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048965-2.5G 048965-500MG
101885-768EA 364.62 USD
101885-768 101885-770
N-(5-Amino-2-methoxyphenyl)cyclobutanecarboxamide
N-(5-Amino-2-methoxyphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD09046190
CAS Number: 926190-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR