N-(3-Amino-2-methylphenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048960-500MG 048960-2.5G
101885-750EA 838.46 USD
101885-750 101885-748
N-(3-Amino-2-methylphenyl)cyclobutanecarboxamide
N-(3-Amino-2-methylphenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD09042828
CAS Number: 926194-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR