N-(3-Aminophenyl)cyclobutanecarboxamide

Supplier: Matrix Scientific

048959-500MG 048959-2.5G
101885-744EA 384.62 USD
101885-744 101885-746
N-(3-Aminophenyl)cyclobutanecarboxamide
N-(3-Aminophenyl)cyclobutanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD09050686
CAS Number: 926264-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR