N-(4-Amino-2-chlorophenyl)cyclopropanecarboxamide
Supplier: Matrix Scientific
048958-2.5G
048958-500MG
101885-740EA
364.62
USD
101885-740
101885-742
N-(4-Amino-2-chlorophenyl)cyclopropanecarboxamide
N-(4-Amino-2-chlorophenyl)cyclopropanecarboxamide
Hazard: Irritant
Formula:
C₁₀H₁₁ClN₂O |
MDL Number:
MFCD09050579 CAS Number: 926247-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...