N-(3-Amino-4-methylphenyl)cyclopropanecarboxamide

Supplier: Matrix Scientific

048954-500MG 048954-2.5G
101885-724EA 364.62 USD
101885-724 101885-726
N-(3-Amino-4-methylphenyl)cyclopropanecarboxamide
N-(3-Amino-4-methylphenyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD09931958
CAS Number: 1016721-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR