N-(4-Amino-2-methylphenyl)-3-methylbutanamide

048951-2.5G
101885-714EA 838.46 USD
101885-714
N-(4-Amino-2-methylphenyl)-3-methylbutanamide
N-(4-Amino-2-methylphenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD03906875
CAS Number: 761409-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR