N-(3-Amino-4-chlorophenyl)-3-methylbutanamide

Supplier: Matrix Scientific

048950-500MG 048950-2.5G
101885-710EA 838.46 USD
101885-710 101885-708
N-(3-Amino-4-chlorophenyl)-3-methylbutanamide
N-(3-Amino-4-chlorophenyl)-3-methylbutanamide

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD09047743
CAS Number: 926222-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR