N-(5-Amino-2-methoxyphenyl)-3-methylbutanamide
Supplier: Matrix Scientific
048948-2.5G
048948-500MG
101885-700EA
166.15
USD
101885-700
101885-702
N-(5-Amino-2-methoxyphenyl)-3-methylbutanamide
N-(5-Amino-2-methoxyphenyl)-3-methylbutanamide
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₂ |
MDL Number:
MFCD09043908 CAS Number: 926194-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...