N-(4-Amino-2-chlorophenyl)-2-methylpropanamide
Supplier: Matrix Scientific
048941-2.5G
048941-500MG
101885-674EA
838.46
USD
101885-674
101885-672
N-(4-Amino-2-chlorophenyl)-2-methylpropanamide
N-(4-Amino-2-chlorophenyl)-2-methylpropanamide
Hazard: Irritant
Formula:
C₁₀H₁₃ClN₂O |
MDL Number:
MFCD03906873 CAS Number: 741271-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...