N-(3-Amino-4-fluorophenyl)-2-methylpropanamide
Supplier: Matrix Scientific
048935-500MG
048935-2.5G
101885-648EA
364.62
USD
101885-648
101885-650
N-(3-Amino-4-fluorophenyl)-2-methylpropanamide
N-(3-Amino-4-fluorophenyl)-2-methylpropanamide
Hazard: Irritant
Formula:
C₁₀H₁₃FN₂O |
MDL Number:
MFCD09046159 CAS Number: 926259-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...