N-(4-Amino-2-chlorophenyl)-3-methoxybenzamide
Supplier: Matrix Scientific
048925-2.5G
048925-500MG
101885-610EA
838.46
USD
101885-610
101885-608
N-(4-Amino-2-chlorophenyl)-3-methoxybenzamide
N-(4-Amino-2-chlorophenyl)-3-methoxybenzamide
Hazard: Irritant
Formula:
C₁₄H₁₃ClN₂O₂ |
MDL Number:
MFCD09047907 CAS Number: 926236-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...