N-(4-Amino-2-methylphenyl)-3-methoxybenzamide

Supplier: Matrix Scientific

048923-500MG 048923-2.5G
101885-600EA 364.62 USD
101885-600 101885-602
N-(4-Amino-2-methylphenyl)-3-methoxybenzamide
N-(4-Amino-2-methylphenyl)-3-methoxybenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD09050944
CAS Number: 926197-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR