N-(3-Aminophenyl)-3-methoxybenzamide

Supplier: Matrix Scientific

048913-500MG 048913-2.5G
101885-560EA 364.62 USD
101885-560 101885-562
N-(3-Aminophenyl)-3-methoxybenzamide
N-(3-Aminophenyl)-3-methoxybenzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MDL Number: MFCD04214420
CAS Number: 354764-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR