N-(3-Amino-4-fluorophenyl)-4-methoxybenzamide

Supplier: Matrix Scientific

048909-500MG 048909-2.5G
101885-544EA 364.62 USD
101885-544 101885-546
N-(3-Amino-4-fluorophenyl)-4-methoxybenzamide
N-(3-Amino-4-fluorophenyl)-4-methoxybenzamide

Hazard: Irritant

Formula: C₁₄H₁₃FN₂O₂
MDL Number: MFCD09049886
CAS Number: 229322-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR