N-(4-Amino-2-methoxyphenyl)tetrahydro-2H-pyran-4-carboxamide

Supplier: Matrix Scientific

048906-2.5G 048906-500MG
101885-532EA 364.62 USD
101885-532 101885-534
N-(4-Amino-2-methoxyphenyl)tetrahydro-2H-pyran-4-carboxamide
N-(4-Amino-2-methoxyphenyl)tetrahydro-2H-pyran-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD13559627
CAS Number: 1220019-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR