N-(4-Amino-2-methylphenyl)tetrahydro-2H-pyran-4-carboxamide

Supplier: Matrix Scientific

048905-2.5G 048905-500MG
101885-530EA 838.46 USD
101885-530 101885-528
N-(4-Amino-2-methylphenyl)tetrahydro-2H-pyran-4-carboxamide
N-(4-Amino-2-methylphenyl)tetrahydro-2H-pyran-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD13559626
CAS Number: 1219949-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR