N-(5-Amino-2-chlorophenyl)tetrahydro-2H-pyran-4-carboxamide

Supplier: Matrix Scientific

048902-500MG 048902-2.5G
101885-516EA 364.62 USD
101885-516 101885-518
N-(5-Amino-2-chlorophenyl)tetrahydro-2H-pyran-4-carboxamide
N-(5-Amino-2-chlorophenyl)tetrahydro-2H-pyran-4-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₂
MDL Number: MFCD13559623
CAS Number: 1220020-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR