N-(4-Amino-2-methoxyphenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048888-2.5G 048888-500MG
101885-460EA 364.62 USD
101885-460 101885-462
N-(4-Amino-2-methoxyphenyl)-2-methoxyacetamide
N-(4-Amino-2-methoxyphenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD09044538
CAS Number: 110506-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR