N-(5-Amino-2-chlorophenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048884-500MG 048884-2.5G
101885-444EA 364.62 USD
101885-444 101885-446
N-(5-Amino-2-chlorophenyl)-2-methoxyacetamide
N-(5-Amino-2-chlorophenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD11568872
CAS Number: 1082188-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR