N-(5-Amino-2-methoxyphenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048883-2.5G 048883-500MG
101885-442EA 838.46 USD
101885-442 101885-440
N-(5-Amino-2-methoxyphenyl)-2-methoxyacetamide
N-(5-Amino-2-methoxyphenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD09042662
CAS Number: 926194-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR