N-(3-Amino-4-fluorophenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048882-2.5G 048882-500MG
101885-436EA 364.62 USD
101885-436 101885-438
N-(3-Amino-4-fluorophenyl)-2-methoxyacetamide
N-(3-Amino-4-fluorophenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₉H₁₁FN₂O₂
MDL Number: MFCD09050758
CAS Number: 926197-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR